Gene Transcripts
- Gene Symbol
- FCHSD2
- Entrez Gene ID
- 9873
- Gene Name
- FCH and double SH3 domains 2
- Gene Aliases
- NWK, SH3MD3
- Chromosome Location
- Chr. 11: 72836743 - 73142682 on Build GRCh38
- UniGene ID
- Hs.744959
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014824.2 |
NP_055639.2
|
15, 16 |
1743 |
|
XM_011545409.1 |
XP_011543711.1
|
15, 16 |
1625 |
|
XM_011545410.2 |
XP_011543712.1
|
15, 16 |
1558 |
|
XM_017018632.1 |
XP_016874121.1
|
15, 16 |
1802 |
|
GenBank mRNA |
AB018312.1 |
- |
14, 15 |
1593 |
|
AK296376.1 |
- |
15, 16 |
1485 |
|
AK297928.1 |
- |
15, 16 |
1627 |
|
AL133567.1 |
- |
10, 11 |
1111 |
|
BC010394.1 |
- |
15, 16 |
1490 |
4091973 |
BC017751.1 |
- |
3, 4 |
293 |
|
View Full Product Details