Gene Transcripts

Gene Symbol
GFPT2
Entrez Gene ID
9945
Gene Name
glutamine-fructose-6-phosphate transaminase 2
Gene Aliases
GFAT, GFAT 2, GFAT2
Chromosome Location
Chr. 5: 180300690 - 180353387 on Build GRCh38
UniGene ID
Hs.696497
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005110.3 NP_005101.1 6 652
XM_006714942.2 XP_006715005.1 6 601
XM_017010100.1 XP_016865589.1 6 599
GenBank mRNA AB016789.1 - 6 581
AK022507.1 - 6 592
AK297750.1 - 4 288
AK314551.1 - 6 592
BC000012.2 - 6 590 3506723
BT009818.1 - 5 482
KU178494.1 - 5 482
KU178495.1 - 5 482

View Full Product Details