Gene Transcripts

Gene Symbol
PDCD6
Entrez Gene ID
10016
Gene Name
programmed cell death 6
Gene Aliases
ALG-2, ALG2, PEF1B
Chromosome Location
Chr. 5: 271621 - 314974 on Build GRCh38
UniGene ID
Hs.50823
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267556.1 NP_001254485.1 5, 6 571
NM_001267557.1 NP_001254486.1 3, 4 373
NM_001267558.1 NP_001254487.1 6, 7 492
NM_013232.3 NP_037364.1 5, 6 577
GenBank mRNA AF035606.1 - 5, 6 603
AK001917.1 - 5, 6 522
AK124302.1 - 1, 2 1832
AK128087.1 - 3458
AK223366.1 - 5, 6 577
AK315370.1 - 5, 6 577
BC012384.1 - 5, 6 545 3859672
BC106706.2 - 5, 6 526 40003290
BC110291.1 - 5, 6 535 5187550
BG390506.1 - 3, 4 303
BI553617.1 - 5, 6 573
BT020072.1 - 5, 6 477
DB495449.1 - 6, 7 477
U58773.1 - 5, 6 508

View Full Product Details