Gene Transcripts
- Gene Symbol
- USP20
- Entrez Gene ID
- 10868
- Gene Name
- ubiquitin specific peptidase 20
- Gene Aliases
- LSFR3A, VDU2, hVDU2
- Chromosome Location
- Chr. 9: 129835417 - 129881838 on Build GRCh38
- UniGene ID
- Hs.5452
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001008563.4 |
NP_001008563.2
|
13, 14 |
1710 |
|
NM_001110303.3 |
NP_001103773.2
|
13, 14 |
1612 |
|
NM_006676.7 |
NP_006667.3
|
13, 14 |
1612 |
|
XM_005251665.3 |
XP_005251722.1
|
13, 14 |
1600 |
|
XM_005251666.3 |
XP_005251723.1
|
8, 9 |
1174 |
|
XM_011518161.2 |
XP_011516463.1
|
14, 15 |
1952 |
|
XM_011518162.2 |
XP_011516464.1
|
13, 14 |
2131 |
|
XM_017014221.1 |
XP_016869710.1
|
8, 9 |
1174 |
|
GenBank mRNA |
AB023220.1 |
- |
12, 13 |
1530 |
|
AY074877.1 |
- |
13, 14 |
1646 |
|
BC039593.1 |
- |
13, 14 |
1556 |
5588842 |
BX462233.2 |
- |
5, 6 |
871 |
|
View Full Product Details