Gene Transcripts
- Gene Symbol
- HHLA2
- Entrez Gene ID
- 11148
- Gene Name
- HERV-H LTR-associating 2
- Gene Aliases
- B7-H5, B7-H7, B7H7, B7y
- Chromosome Location
- Chr. 3: 108296490 - 108378285 on Build GRCh38
- UniGene ID
- Hs.225968
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001282556.1 |
NP_001269485.1
|
4, 5 |
629 |
|
NM_001282557.1 |
NP_001269486.1
|
4, 5 |
560 |
|
NM_001282558.1 |
NP_001269487.1
|
5, 6 |
707 |
|
NM_001282559.1 |
NP_001269488.1
|
5, 6 |
445 |
|
NM_007072.3 |
NP_009003.1
|
4, 5 |
820 |
|
XM_005247080.3 |
XP_005247137.1
|
5, 6 |
690 |
|
XM_011512362.1 |
XP_011510664.1
|
5, 6 |
690 |
|
XM_011512363.1 |
XP_011510665.1
|
4, 5 |
620 |
|
XM_011512364.1 |
XP_011510666.1
|
4, 5 |
571 |
|
XM_011512367.2 |
XP_011510669.1
|
5, 6 |
690 |
|
XM_017005642.1 |
XP_016861131.1
|
4, 5 |
620 |
|
GenBank mRNA |
AF126162.1 |
- |
4, 5 |
560 |
|
AK000692.1 |
- |
3, 4 |
555 |
|
AK027132.1 |
- |
1, 2 |
113 |
|
AK296644.1 |
- |
5, 6 |
388 |
|
AK315109.1 |
- |
5, 6 |
652 |
|
BC035971.1 |
- |
4, 5 |
820 |
4606764 |
View Full Product Details