Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014899.3 | NP_055714.3 | 12 | 2354 | |
XM_011543279.2 | XP_011541581.1 | 11 | 2068 | ||
XM_017009236.1 | XP_016864725.1 | 13 | 2115 | ||
XM_017009237.1 | XP_016864726.1 | 13 | 2236 | ||
GenBank mRNA | AB020685.1 | - | 12 | 2152 | |
AK290034.1 | - | 12 | 2127 | ||
BC041337.2 | - | 12 | 2159 | 5285051 |