Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012228.3 | NP_036360.3 | 5 | 627 | |
GenBank mRNA | AF122004.1 | - | 5 | 544 | |
AF151889.1 | - | 5 | 575 | ||
BC018030.1 | - | 5 | 622 | 4799583 | |
BC117471.1 | - | 5 | 676 | 40126022 | |
BC130380.1 | - | 5 | 676 | 40146411 |