Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014611.2 | NP_055426.1 | 65 | 11153 | |
XM_005248700.3 | XP_005248757.1 | 26 | 5085 | ||
XM_006715405.3 | XP_006715468.1 | 29 | 6631 | ||
XM_011535635.2 | XP_011533937.1 | 65 | 10958 | ||
XM_011535636.2 | XP_011533938.1 | 65 | 11123 | ||
GenBank mRNA | AB002299.2 | - | 19 | 3809 | |
AF503925.1 | - | 65 | 11082 | ||
AK125181.1 | - | 3 | 953 | ||
CR627399.1 | - | 11 | 2608 |