Gene Transcripts

Gene Symbol
CLEC5A
Entrez Gene ID
23601
Gene Name
C-type lectin domain family 5 member A
Gene Aliases
CLECSF5, MDL-1, MDL1
Chromosome Location
Chr. 7: 141927356 - 141947009 on Build GRCh38
UniGene ID
Hs.446235
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301167.1 NP_001288096.1 2 240
NM_013252.2 NP_037384.1 2 216
XM_011515995.2 XP_011514297.1 2 232
XM_017011915.1 XP_016867404.1 2 234
XM_017011916.1 XP_016867405.1 2 242
XM_017011917.1 XP_016867406.1 2 242
GenBank mRNA AF139768.1 - 2 170
AJ271684.1 - 2 216
AK292766.1 - 2 205
AK295200.1 - 2 177
AK307095.1 - 2 240
AK313703.1 - 2 205
BC093714.1 - 1 40 7939559
BC112099.1 - 1 40 8327567
BC113098.1 - 2 158 40076723
BC113099.1 - 2 158 40076725
BC143392.1 - 1 40

View Full Product Details