Gene Transcripts
- Gene Symbol
- TTLL1
- Entrez Gene ID
- 25809
- Gene Name
- tubulin tyrosine ligase like 1
- Gene Aliases
- C22orf7, HS323M22B
- Chromosome Location
- Chr. 22: 43039516 - 43089428 on Build GRCh38
- UniGene ID
- Hs.660298
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012263.4 |
NP_036395.1
|
5, 6 |
734 |
|
NR_027779.1 |
|
6, 7 |
838 |
|
XM_006724225.3 |
XP_006724288.1
|
5, 6 |
700 |
|
XM_011530108.2 |
XP_011528410.1
|
6, 7 |
761 |
|
XM_011530110.2 |
XP_011528412.1
|
5, 6 |
721 |
|
XM_017028751.1 |
XP_016884240.1
|
6, 7 |
736 |
|
XM_017028752.1 |
XP_016884241.1
|
5, 6 |
647 |
|
XM_017028753.1 |
XP_016884242.1
|
6, 7 |
836 |
|
GenBank mRNA |
AF104927.1 |
- |
5, 6 |
635 |
|
AF173935.1 |
- |
6, 7 |
748 |
|
AK095996.1 |
- |
5, 6 |
710 |
|
AK315658.1 |
- |
5, 6 |
659 |
|
AL096883.1 |
- |
5, 6 |
654 |
|
AL096886.1 |
- |
4, 5 |
582 |
|
AL136687.1 |
- |
5, 6 |
734 |
|
AL589867.1 |
- |
3, 4 |
493 |
|
BC014968.1 |
- |
5, 6 |
721 |
4876418 |
CR456599.1 |
- |
3, 4 |
497 |
|
View Full Product Details