Gene Transcripts

Gene Symbol
FBXW8
Entrez Gene ID
26259
Gene Name
F-box and WD repeat domain containing 8
Gene Aliases
FBW6, FBW8, FBX29, FBXO29, FBXW6
Chromosome Location
Chr. 12: 116910949 - 117032498 on Build GRCh38
UniGene ID
Hs.624537
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012174.1 NP_036306.1 6 757
NM_153348.2 NP_699179.2 6 955
XM_005253867.2 XP_005253924.1 5 813
XM_005253868.3 XP_005253925.1 5 646
XM_017019175.1 XP_016874664.1 6 732
XM_017019176.1 XP_016874665.1 7 964
XM_017019177.1 XP_016874666.1 7 964
XM_017019178.1 XP_016874667.1 2 1706
GenBank mRNA AF176707.1 - 6 733
AK289621.1 - 6 748
BC037296.2 - 6 955 4821240

Gene Symbol
LOC100506551
Entrez Gene ID
100506551
Gene Name
uncharacterized LOC100506551
Gene Aliases
Chromosome Location
Chr. 12: 116977440 - 116987337 on Build GRCh38
UniGene ID
Hs.657861
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_103809.1 2 1814
GenBank mRNA AK127077.1 - 2 1814

View Full Product Details