Gene Transcripts

Gene Symbol
COQ4
Entrez Gene ID
51117
Gene Name
coenzyme Q4
Gene Aliases
CGI-92, COQ10D7
Chromosome Location
Chr. 9: 128322486 - 128334072 on Build GRCh38
UniGene ID
Hs.98541
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001305942.1 NP_001292871.1 4 672
NM_016035.4 NP_057119.2 7 993
GenBank mRNA AF151850.1 - 7 964
AK025376.1 - 7 907
AK095083.1 - 3 2458
AK128853.1 - 2 4657
AK313650.1 - 7 976
BC011895.2 - 7 663 3944239
CB111694.1 - 4 343
EU216419.1 - 7 975
EU216420.1 - 7 893
EU216421.1 - 7 727
EU216422.1 - 7 665
EU216423.1 - 6 589

View Full Product Details