Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016100.4 | NP_057184.1 | 3 | 226 | |
NM_181527.3 | NP_852668.1 | 3 | 313 | ||
NM_181528.3 | NP_852669.1 | 3 | 226 | ||
GenBank mRNA | AF085355.1 | - | 3 | 189 | |
AK311819.1 | - | 3 | 179 | ||
AL136641.1 | - | 3 | 158 | ||
BC005181.1 | - | 3 | 154 | 3950192 | |
BC008446.1 | - | 3 | 175 | 4104421 | |
BG548527.1 | - | 3 | 174 | ||
BX338359.1 | - | 4 | 796 | ||
CA867744.1 | - | 3 | 400 | ||
CB959658.1 | - | 3 | 250 |