Gene Transcripts

Gene Symbol
TRIM33
Entrez Gene ID
51592
Gene Name
tripartite motif containing 33
Gene Aliases
ECTO, PTC7, RFG7, TF1G, TIF1G, TIF1GAMMA, TIFGAMMA
Chromosome Location
Chr. 1: 114392777 - 114511160 on Build GRCh38
UniGene ID
Hs.26837
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015906.3 NP_056990.3 18 3091
NM_033020.2 NP_148980.2 18 3091
XM_005270936.3 XP_005270993.1 19 3084
XM_005270937.3 XP_005270994.1 19 3084
XM_011541568.2 XP_011539870.1 19 3081
XM_017001452.1 XP_016856941.1 16 2366
XM_017001453.1 XP_016856942.1 16 2366
GenBank mRNA AB029036.1 - 18 3020
AF119043.1 - 18 3091
AF220136.1 - 18 3091
AF220137.1 - 18 3091
AJ132948.1 - 18 2782
AK023434.1 - 16 2304
AK057487.1 - 13 1308
AK301997.1 - 13 1769

View Full Product Details