Gene Transcripts

Gene Symbol
MRPL48
Entrez Gene ID
51642
Gene Name
mitochondrial ribosomal protein L48
Gene Aliases
CGI-118, HSPC290, L48MT, MRP-L48
Chromosome Location
Chr. 11: 73787872 - 73864611 on Build GRCh38
UniGene ID
Hs.503239
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318498.1 NP_001305427.1 7, 8 622
NM_001318499.1 NP_001305428.1 7, 8 687
NM_016055.5 NP_057139.1 6, 7 564
NR_134658.1 5, 6 526
NR_134659.1 8, 9 817
NR_134660.1 6, 7 682
XM_017017919.1 XP_016873408.1 7, 8 657
GenBank mRNA AF151876.1 - 6, 7 1120
AF161408.1 - 6, 7 567
AK297746.1 - 7, 8 622
AK310005.1 - 8, 9 796
BC009764.1 - 6, 7 557 4093230
BI549412.1 - 5, 6 518
BM817807.1 - 6, 7 562
BM828996.1 - 6, 7 563
BQ446725.1 - 2, 3 430
BX354271.2 - 6, 7 647
CF529680.1 - 2, 3 430

View Full Product Details