Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018946.3 | NP_061819.2 | 2 | 449 | |
XM_011518787.2 | XP_011517089.1 | 1 | 42 | ||
XM_017014811.1 | XP_016870300.1 | 1 | 19 | ||
GenBank mRNA | AF257466.1 | - | 2 | 317 | |
AK001659.1 | - | 2 | 387 | ||
AK316608.1 | - | 2 | 409 | ||
AL526281.3 | - | 2 | 449 | ||
BC000008.2 | - | 2 | 395 | 3505324 | |
BC019315.1 | - | 2 | 377 | 2821389 | |
FJ224349.1 | - | 2 | 317 |