Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021027.2 | NP_066307.1 | 1, 2 | 884 | |
GenBank mRNA | AF056188.1 | - | 1, 2 | 865 | |
AK313510.1 | - | 1, 2 | 923 | ||
AY435144.1 | - | 1, 2 | 847 | ||
BC058844.1 | - | 1, 2 | 890 | 4593613 | |
DQ364246.1 | - | 1, 2 | 199 |