Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001307977.1 | NP_001294906.1 | 4 | 418 | |
NM_018093.3 | NP_060563.2 | 5 | 497 | ||
XM_005274055.2 | XP_005274112.1 | 5 | 2358 | ||
GenBank mRNA | AK001301.1 | - | 5 | 497 | |
AK292330.1 | - | 5 | 867 | ||
AK303186.1 | - | 4 | 339 | ||
AY500828.1 | - | 4 | 349 | ||
BC006351.2 | - | 4 | 340 | 4111690 |