Gene Transcripts

Gene Symbol
TEX10
Entrez Gene ID
54881
Gene Name
testis expressed 10
Gene Aliases
Ipi1, bA208F1.2
Chromosome Location
Chr. 9: 100302075 - 100352977 on Build GRCh38
UniGene ID
Hs.494648
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161584.1 NP_001155056.1 4 1090
NM_017746.3 NP_060216.2 4 1146
XM_011518798.1 XP_011517100.1 4 1108
XM_017014848.1 XP_016870337.1 4 635
GenBank mRNA AB060968.1 - 4 1062
AK000294.1 - 4 1062
AK298916.1 - 6 669
AK302619.1 - 4 1093
AK303662.1 - 4 586
AK304869.1 - 3 863
AY598337.1 - 3 931
BC030652.2 - 4 982 5268105

View Full Product Details