Gene Transcripts

Gene Symbol
TEX10
Entrez Gene ID
54881
Gene Name
testis expressed 10
Gene Aliases
Ipi1, bA208F1.2
Chromosome Location
Chr. 9: 100302075 - 100352977 on Build GRCh38
UniGene ID
Hs.494648
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161584.1 NP_001155056.1 15 2876
NM_017746.3 NP_060216.2 15 2989
XM_011518798.1 XP_011517100.1 13 2576
XM_017014848.1 XP_016870337.1 15 2478
GenBank mRNA AA436854.1 - 1 127
AB060968.1 - 15 2905
AJ420436.1 - 10 1414
AK000294.1 - 15 2905
AK091669.1 - 2 2239
AK298916.1 - 17 2512
AK302619.1 - 15 2879
AY598337.1 - 14 2774
BC030652.2 - 15 2825 5268105
BF981005.1 - 3 498

View Full Product Details