Gene Transcripts

Gene Symbol
C17orf80
Entrez Gene ID
55028
Gene Name
chromosome 17 open reading frame 80
Gene Aliases
HLC-8, MIG3, SPEP1
Chromosome Location
Chr. 17: 73232233 - 73248959 on Build GRCh38
UniGene ID
Hs.12929
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100621.2 NP_001094091.1 3 1658
NM_001100622.2 NP_001094092.1 3 1658
NM_001288770.1 NP_001275699.1 3 1819
NM_001288771.1 NP_001275700.1 3 1819
NM_017941.5 NP_060411.2 3 1658
XM_005257487.3 XP_005257544.1 3 1651
XM_006721966.3 XP_006722029.1 3 1658
XM_011524961.1 XP_011523263.1 3 1658
XM_011524962.2 XP_011523264.1 2 1556
XM_017024806.1 XP_016880295.1 2 1584
GenBank mRNA AK000728.1 - 3 1617
AK292838.1 - 3 1587
AK301905.1 - 3 1819
AL136740.1 - 3 1625
AY163812.1 - 3 1643
AY239293.1 - 1 328
BC005005.2 - 2 1563 3544728

View Full Product Details