Gene Transcripts

Gene Symbol
TRIM68
Entrez Gene ID
55128
Gene Name
tripartite motif containing 68
Gene Aliases
GC109, RNF137, SS-56, SS56
Chromosome Location
Chr. 11: 4598672 - 4608259 on Build GRCh38
UniGene ID
Hs.523438
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304496.1 NP_001291425.1 6 841
NM_018073.7 NP_060543.5 7 1324
GenBank mRNA AF360739.1 - 7 1277
AF439153.1 - 6 1034
AK001231.1 - 4 529
AK022923.1 - 4 494
AK056523.1 - 7 1286
AK291166.1 - 7 1236
AK301120.1 - 5 556
AL832730.1 - 7 1274
AY005802.1 - 6 710
BC075058.2 - 6 1058 30915367
BC109063.2 - 7 1243 40033795

View Full Product Details