Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018141.3 | NP_060611.2 | 7 | 614 | |
XM_005249206.2 | XP_005249263.1 | 7 | 623 | ||
XM_011514724.2 | XP_011513026.1 | 8 | 618 | ||
XM_011514725.2 | XP_011513027.1 | 8 | 610 | ||
XM_017010996.1 | XP_016866485.1 | 7 | 615 | ||
GenBank mRNA | AF113220.1 | - | 7 | 574 | |
AK001429.1 | - | 7 | 583 | ||
AK098446.1 | - | 7 | 577 | ||
AK298219.1 | - | 7 | 597 | ||
AK316599.1 | - | 7 | 580 | ||
AW974308.1 | - | 1 | 307 | ||
BC012560.1 | - | 7 | 584 | 4277803 | |
BG776014.1 | - | 7 | 617 |