Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018409.3 | NP_060879.2 | 5 | 1312 | |
XM_005263125.4 | XP_005263182.1 | 4 | 592 | ||
XM_006714260.3 | XP_006714323.1 | 6 | 1642 | ||
XM_006714265.3 | XP_006714328.1 | 5 | 702 | ||
XM_011532110.2 | XP_011530412.1 | 5 | 1515 | ||
XM_011532111.2 | XP_011530413.1 | 5 | 565 | ||
XM_017008410.1 | XP_016863899.1 | 6 | 2525 | ||
XM_017008411.1 | XP_016863900.1 | 5 | 1130 | ||
GenBank mRNA | AB037746.1 | - | 5 | 1312 | |
AK126913.1 | - | 6 | 1643 | ||
AL161975.1 | - | 4 | 2406 | ||
BC150161.1 | - | 5 | 819 | 40126591 | |
BC152444.1 | - | 5 | 1312 | 8862572 |