Gene Transcripts

Gene Symbol
UBFD1
Entrez Gene ID
56061
Gene Name
ubiquitin family domain containing 1
Gene Aliases
UBPH
Chromosome Location
Chr. 16: 23557535 - 23574393 on Build GRCh38
UniGene ID
Hs.3459
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019116.2 NP_061989.2 7 1070
XM_011545894.2 XP_011544196.1 6 993
GenBank mRNA AK092696.1 - 7 884
AK093029.1 - 2 2154
AK096189.1 - 3 151
AK124139.1 - 8 846

View Full Product Details