Gene Transcripts

Gene Symbol
GRHL3
Entrez Gene ID
57822
Gene Name
grainyhead like transcription factor 3
Gene Aliases
SOM, TFCP2L4, VWS2
Chromosome Location
Chr. 1: 24319322 - 24364482 on Build GRCh38
UniGene ID
Hs.657920
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195010.1 NP_001181939.1 2, 3 380
NM_021180.3 NP_067003.2 2, 3 276
NM_198173.2 NP_937816.1 2, 3 445
NM_198174.2 NP_937817.3 2, 3 445
XM_011541869.1 XP_011540171.1 2, 3 326
XM_011541870.2 XP_011540172.1 1, 2 247
GenBank mRNA AK074386.1 - 2, 3 342
AK315164.1 - 2, 3 247
AY231160.1 - 2, 3 276
AY231161.1 - 1, 2 89
BC036890.2 - 2, 3 391 5502632
BU848928.1 - 2, 3 376
DA714132.1 - 2, 3 445

View Full Product Details