Gene Transcripts
- Gene Symbol
- ERVK-10
- Entrez Gene ID
- 100616101
- Gene Name
- endogenous retrovirus group K member 10
- Gene Aliases
- PR, Protease, Proteinase
- Chromosome Location
- Chr. : - on Build
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
CN345079.1 |
- |
1 |
546 |
|
- Gene Symbol
- LOC105374013
- Entrez Gene ID
- 105374013
- Gene Name
- endogenous retrovirus group K member 5 Gag polyprotein
- Gene Aliases
- Chromosome Location
- Chr. 3: 101686870 - 101696682 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- LOC107986062
- Entrez Gene ID
- 107986062
- Gene Name
- endogenous retrovirus group K member 9 Pol protein-like
- Gene Aliases
- Chromosome Location
- Chr. 3: 9848661 - 9854308 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- LOC107986113
- Entrez Gene ID
- 107986113
- Gene Name
- endogenous retrovirus group K member 7 Gag polyprotein
- Gene Aliases
- Chromosome Location
- Chr. 3: 113029918 - 113033146 on Build GRCh38
- Species
- Homo sapiens
View Full Product Details