Gene Transcripts

Gene Symbol
MLST8
Entrez Gene ID
64223
Gene Name
MTOR associated protein, LST8 homolog
Gene Aliases
GBL, GbetaL, LST8, POP3, WAT1
Chromosome Location
Chr. 16: 2205177 - 2209417 on Build GRCh38
UniGene ID
Hs.29203
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199173.1 NP_001186102.1 8 1086
NM_001199174.1 NP_001186103.1 8 925
NM_001199175.1 NP_001186104.1 8 1113
NM_022372.4 NP_071767.3 8 1116
XM_005255475.3 XP_005255532.2 8 892
XM_005255478.2 XP_005255535.1 8 1097
XM_005255479.2 XP_005255536.1 8 1024
XM_017023548.1 XP_016879037.1 8 1081
XM_017023549.1 XP_016879038.1 8 822
XM_017023551.1 XP_016879040.1 8 1131
GenBank mRNA AF195883.1 - 5 437
AK021536.1 - 8 1085
AK022227.1 - 7 770
AK098762.1 - 8 1021
AK123195.1 - 1 891
AK302201.1 - 7 925
AY223837.1 - 7 926
BC001313.2 - 8 806 3454025
BC017119.1 - 7 817 3910883
BC052292.1 - 7 1340 5926090
BC088354.1 - 8 834 6168345

View Full Product Details