Gene Transcripts

Gene Symbol
NOC3L
Entrez Gene ID
64318
Gene Name
NOC3 like DNA replication regulator
Gene Aliases
AD24, C10orf117, FAD24
Chromosome Location
Chr. 10: 94333226 - 94362976 on Build GRCh38
UniGene ID
Hs.74899
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022451.10 NP_071896.8 12, 13 1615
XM_017016544.1 XP_016872033.1 10, 11 1668
GenBank mRNA AB077992.1 - 12, 13 1502
AK022882.1 - 4, 5 526
AK026953.1 - 2, 3 226
AK091246.1 - 12, 13 1572
AK292520.1 - 12, 13 1597
AK302028.1 - 6, 7 700
BC049850.1 - 12, 13 1565 5549517

View Full Product Details