Gene Transcripts

Gene Symbol
MARC1
Entrez Gene ID
64757
Gene Name
mitochondrial amidoxime reducing component 1
Gene Aliases
MOSC1
Chromosome Location
Chr. 1: 220786697 - 220814399 on Build GRCh38
UniGene ID
Hs.497816
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022746.3 NP_073583.3 3 819
XM_011509900.2 XP_011508202.1 3 819
XM_011509903.2 XP_011508205.1 3 1195
XM_011509904.2 XP_011508206.1 3 472
XM_017002096.1 XP_016857585.1 3 399
XM_017002097.1 XP_016857586.1 3 452
GenBank mRNA AK026043.1 - 3 615
AK092439.1 - 2 1290
AK094105.1 - 4 482
AK225416.1 - 3 615
AK290812.1 - 3 591
BC010619.1 - 3 587 3872779

View Full Product Details