Gene Transcripts

Gene Symbol
CHMP6
Entrez Gene ID
79643
Gene Name
charged multivesicular body protein 6
Gene Aliases
VPS20
Chromosome Location
Chr. 17: 80991841 - 81000133 on Build GRCh38
UniGene ID
Hs.514560
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024591.4 NP_078867.2 6, 7 565
XM_005257668.1 XP_005257725.1 6, 7 565
GenBank mRNA AK021811.1 - 6, 7 503
AK223187.1 - 6, 7 527
AK292105.1 - 6, 7 531
AY329087.1 - 6, 7 487
BC010108.2 - 6, 7 511 3604039
BP196406.1 - 565
CR457284.1 - 6, 7 487

View Full Product Details