Gene Transcripts

Gene Symbol
NAA40
Entrez Gene ID
79829
Gene Name
N(alpha)-acetyltransferase 40, NatD catalytic subunit
Gene Aliases
NAT11, PATT1
Chromosome Location
Chr. 11: 63938970 - 63957322 on Build GRCh38
UniGene ID
Hs.523753
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300800.1 NP_001287729.1 3, 4 363
NM_024771.3 NP_079047.2 3, 4 292
XM_006718689.3 XP_006718752.1 3, 4 279
XM_011545254.2 XP_011543556.1 4, 5 366
XM_011545255.2 XP_011543557.1 4, 5 366
GenBank mRNA AK023910.1 - 3, 4 261
AK299041.1 - 3, 4 357
AK300451.1 - 2, 3 213
AK303557.1 - 2, 3 213
BC041617.1 - 3, 4 221 5921480
BC052298.1 - 3, 4 221 6292760
BX647309.1 - 2, 3 1318
DA649381.1 - 3, 4 297
DC307984.1 - 3, 4 446
HY009487.1 - 3, 4 363
JF263566.1 - 3, 4 261

View Full Product Details