Gene Transcripts

Gene Symbol
C10orf88
Entrez Gene ID
80007
Gene Name
chromosome 10 open reading frame 88
Gene Aliases
Chromosome Location
Chr. 10: 122930903 - 122954403 on Build GRCh38
UniGene ID
Hs.501286
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024942.3 NP_079218.2 5 960
GenBank mRNA AK023552.1 - 5 816
BC032645.1 - 5 800 5519534

View Full Product Details