Gene Transcripts
- Gene Symbol
- CFAP43
- Entrez Gene ID
- 80217
- Gene Name
- cilia and flagella associated protein 43
- Gene Aliases
- C10orf79, WDR96, bA373N18.2
- Chromosome Location
- Chr. 10: 104129888 - 104232363 on Build GRCh38
- UniGene ID
- Hs.288927
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_025145.5 |
NP_079421.5
|
25, 26 |
3447 |
|
XM_005270171.2 |
XP_005270228.1
|
25, 26 |
3451 |
|
XM_005270172.3 |
XP_005270229.1
|
25, 26 |
3451 |
|
XM_011540196.2 |
XP_011538498.1
|
26, 27 |
3565 |
|
XM_011540197.2 |
XP_011538499.1
|
26, 27 |
3565 |
|
XM_011540198.2 |
XP_011538500.1
|
25, 26 |
3448 |
|
XM_011540199.2 |
XP_011538501.1
|
25, 26 |
3448 |
|
XM_011540200.2 |
XP_011538502.1
|
26, 27 |
3565 |
|
XM_011540201.2 |
XP_011538503.1
|
26, 27 |
3565 |
|
XM_011540202.2 |
XP_011538504.1
|
22, 23 |
2881 |
|
XM_011540203.2 |
XP_011538505.1
|
24, 25 |
3123 |
|
XM_017016681.1 |
XP_016872170.1
|
26, 27 |
3562 |
|
XM_017016682.1 |
XP_016872171.1
|
23, 24 |
3217 |
|
XM_017016683.1 |
XP_016872172.1
|
23, 24 |
3003 |
|
GenBank mRNA |
AL833836.1 |
- |
11, 12 |
1413 |
|
View Full Product Details