Gene Transcripts

Gene Symbol
PAAF1
Entrez Gene ID
80227
Gene Name
proteasomal ATPase associated factor 1
Gene Aliases
PAAF, Rpn14, WDR71
Chromosome Location
Chr. 11: 73876699 - 73927736 on Build GRCh38
UniGene ID
Hs.525017
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267803.1 NP_001254732.1 8 696
NM_001267804.1 NP_001254733.1 8 687
NM_001267805.1 NP_001254734.1 8 683
NM_001267806.1 NP_001254735.1 7 570
NM_025155.2 NP_079431.1 7 627
XM_005274308.2 XP_005274365.2 5 1219
XM_005274309.1 XP_005274366.1 6 486
XM_017018357.1 XP_016873846.1 8 615
XM_017018358.1 XP_016873847.1 8 620
XM_017018359.1 XP_016873848.1 5 1178
GenBank mRNA AF087895.1 - 8 621
AK021910.1 - 7 608
AK222501.1 - 7 608
AK225520.1 - 7 595
AK298258.1 - 7 570
AK309133.1 - 5 834
AK316394.1 - 7 841
BC006142.2 - 7 580 3937356
BC021541.1 - 7 591 3942680
BC028628.1 - 8 661 5263875
CD243380.1 - 8 673

View Full Product Details