Gene Transcripts

Gene Symbol
LY6G6C
Entrez Gene ID
80740
Gene Name
lymphocyte antigen 6 complex, locus G6C
Gene Aliases
C6orf24, G6c, NG24
Chromosome Location
Chr. 6: 31718648 - 31721734 on Build GRCh38
UniGene ID
Hs.732272
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025261.2 NP_079537.1 2 200
GenBank mRNA AJ315533.1 - 2 145
AJ315534.1 - 3 256
AY359119.1 - 2 200
BC036302.1 - 2 175 4776231

View Full Product Details