Gene Transcripts

Gene Symbol
RSPH6A
Entrez Gene ID
81492
Gene Name
radial spoke head 6 homolog A
Gene Aliases
RSHL1, RSP4, RSP6, RSPH4B
Chromosome Location
Chr. 19: 45795710 - 45815347 on Build GRCh38
UniGene ID
Hs.146544
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030785.3 NP_110412.1 2 888
XM_011527351.1 XP_011525653.1 2 864
GenBank mRNA AK093068.1 - 1 158
AK098837.1 - 2 833
AK223347.1 - 2 833
AL136761.1 - 2 860
BC057785.1 - 2 716
BC136351.1 - 2 769 9020338
BC136352.1 - 2 769 9020339

View Full Product Details