Gene Transcripts

Gene Symbol
ZMYND12
Entrez Gene ID
84217
Gene Name
zinc finger MYND-type containing 12
Gene Aliases
Chromosome Location
Chr. 1: 42430329 - 42456267 on Build GRCh38
UniGene ID
Hs.294009
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146192.1 NP_001139664.1 3 641
NM_032257.4 NP_115633.3 4 813
GenBank mRNA AK057384.1 - 4 813
AK301839.1 - 3 627
AL136858.1 - 4 620
BC024186.2 - 4 802 4826255
BQ441452.1 - 1 102

View Full Product Details