Gene Transcripts
- Gene Symbol
- ATAD1
- Entrez Gene ID
- 84896
- Gene Name
- ATPase family, AAA domain containing 1
- Gene Aliases
- AFDC1, FNP001, THORASE
- Chromosome Location
- Chr. 10: 87751512 - 87841345 on Build GRCh38
- UniGene ID
- Hs.435948
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001321967.1 |
NP_001308896.1
|
3, 4 |
346 |
|
NM_001321968.1 |
NP_001308897.1
|
3, 4 |
346 |
|
NM_001321969.1 |
NP_001308898.1
|
3, 4 |
346 |
|
NM_032810.3 |
NP_116199.2
|
3, 4 |
718 |
|
NR_135914.1 |
|
3, 4 |
346 |
|
XM_005270252.4 |
XP_005270309.1
|
3, 4 |
440 |
|
XM_011540302.1 |
XP_011538604.1
|
1, 2 |
111 |
|
XM_011540303.2 |
XP_011538605.1
|
3, 4 |
393 |
|
XM_017016847.1 |
XP_016872336.1
|
3, 4 |
587 |
|
XM_017016848.1 |
XP_016872337.1
|
3, 4 |
680 |
|
XM_017016849.1 |
XP_016872338.1
|
3, 4 |
469 |
|
XM_017016850.1 |
XP_016872339.1
|
3, 4 |
520 |
|
XM_017016851.1 |
XP_016872340.1
|
3, 4 |
393 |
|
GenBank mRNA |
AF361493.1 |
- |
2, 3 |
552 |
|
AK075223.1 |
- |
3, 4 |
647 |
|
AK304297.1 |
- |
1, 2 |
109 |
|
AL834370.1 |
- |
2, 3 |
131 |
|
BC063530.1 |
- |
2, 3 |
177 |
|
BC073998.1 |
- |
2, 3 |
289 |
6199813 |
View Full Product Details