Gene Transcripts

Gene Symbol
MAEL
Entrez Gene ID
84944
Gene Name
maelstrom spermatogenic transposon silencer
Gene Aliases
CT128, SPATA35
Chromosome Location
Chr. 1: 166921121 - 167022214 on Build GRCh38
UniGene ID
Hs.651245
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286377.1 NP_001273306.1 9, 10 1219
NM_001286378.1 NP_001273307.1 11, 12 1085
NM_032858.2 NP_116247.1 10, 11 1312
XM_006711583.1 XP_006711646.1 11, 12 1116
XM_011510067.1 XP_011508369.1 10, 11 1278
XM_011510068.1 XP_011508370.1 11, 12 1069
XM_017002602.1 XP_016858091.1 12, 13 1453
XM_017002603.1 XP_016858092.1 11, 12 1115
GenBank mRNA AK027810.1 - 10, 11 1119
AK131066.1 - 10, 11 1122
AK302253.1 - 9, 10 1035
AL133073.1 - 9, 10 876
BC028595.2 - 10, 11 1092 5270252
BC034310.1 - 10, 11 1740
DQ076156.2 - 10, 11 1121
GU383113.1 - 9, 10 1028
GU383114.1 - 11, 12 1085

View Full Product Details