Gene Transcripts

Gene Symbol
RFT1
Entrez Gene ID
91869
Gene Name
RFT1 homolog
Gene Aliases
CDG1N
Chromosome Location
Chr. 3: 53071151 - 53130469 on Build GRCh38
UniGene ID
Hs.631910
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052859.3 NP_443091.1 12 1351
XM_011534216.2 XP_011532518.1 12 1449
XM_017007460.1 XP_016862949.1 12 1363
XM_017007461.1 XP_016862950.1 13 1535
GenBank mRNA AJ318099.1 - 12 1335
BC006846.1 - 3 265
BC043595.1 - 12 1344 5769824

View Full Product Details