Gene Transcripts

Gene Symbol
UBTD2
Entrez Gene ID
92181
Gene Name
ubiquitin domain containing 2
Gene Aliases
DCUBP
Chromosome Location
Chr. 5: 172209645 - 172283797 on Build GRCh38
UniGene ID
Hs.131570
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152277.2 NP_689490.2 2 336
XM_017010022.1 XP_016865511.1 2 346
XM_017010023.1 XP_016865512.1 2 270
GenBank mRNA AK313274.1 - 2 302
BC019910.1 - 2 184 4402070
BU849695.1 - 2 351

View Full Product Details