Gene Transcripts

Gene Symbol
CALHM3
Entrez Gene ID
119395
Gene Name
calcium homeostasis modulator 3
Gene Aliases
FAM26A, bA225H22.7
Chromosome Location
Chr. 10: 103472804 - 103479240 on Build GRCh38
UniGene ID
Hs.364624
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001129742.1 NP_001123214.1 3 850
GenBank mRNA BC039499.1 - 2 226
BC043367.1 - 3 850 5110383

View Full Product Details