Gene Transcripts

Gene Symbol
LYZL4
Entrez Gene ID
131375
Gene Name
lysozyme like 4
Gene Aliases
LYC4, LYZA
Chromosome Location
Chr. 3: 42360767 - 42427911 on Build GRCh38
UniGene ID
Hs.234895
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304386.1 NP_001291315.1 2 304
NM_144634.3 NP_653235.1 2 323
XM_011533355.2 XP_011531657.1 2 366
XM_017005705.1 XP_016861194.1 4 678
XM_017005706.1 XP_016861195.1 3 436
GenBank mRNA AF326749.1 - 2 252
BC016747.2 - 2 294 4830396
BX282166.1 - 2 256
HY037577.1 - 2 307

View Full Product Details