Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001177316.1 | NP_001170787.1 | 7 | 818 | |
NM_001177317.1 | NP_001170788.1 | 7 | 821 | ||
NM_080877.2 | NP_543153.1 | 7 | 783 | ||
XM_011518257.2 | XP_011516559.1 | 7 | 759 | ||
XM_011518258.1 | XP_011516560.1 | 6 | 649 | ||
XM_011518259.1 | XP_011516561.1 | 7 | 783 | ||
XM_011518260.1 | XP_011516562.1 | 7 | 701 | ||
XM_011518261.2 | XP_011516563.1 | 7 | 774 | ||
XM_017014290.1 | XP_016869779.1 | 8 | 869 | ||
XM_017014291.1 | XP_016869780.1 | 7 | 759 | ||
XM_017014292.1 | XP_016869781.1 | 7 | 759 | ||
GenBank mRNA | AB055000.1 | - | 7 | 667 | |
AK095999.1 | - | 7 | 783 | ||
AL832449.1 | - | 7 | 1829 | ||
BC029942.1 | - | 7 | 821 |