Gene Transcripts
- Gene Symbol
- RNF38
- Entrez Gene ID
- 152006
- Gene Name
- ring finger protein 38
- Gene Aliases
- Chromosome Location
- Chr. 9: 36336398 - 36487384 on Build GRCh38
- UniGene ID
- Hs.333503
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022781.4 |
NP_073618.3
|
6, 7 |
1089 |
|
NM_194328.2 |
NP_919309.1
|
6, 7 |
1256 |
|
NM_194329.2 |
NP_919310.1
|
5, 6 |
939 |
|
NM_194330.2 |
NP_919311.1
|
6, 7 |
987 |
|
NM_194332.2 |
NP_919313.1
|
5, 6 |
1106 |
|
XM_005251366.3 |
XP_005251423.1
|
6, 7 |
1093 |
|
XM_005251367.3 |
XP_005251424.1
|
6, 7 |
991 |
|
XM_005251368.3 |
XP_005251425.1
|
6, 7 |
1105 |
|
XM_006716721.3 |
XP_006716784.1
|
5, 6 |
953 |
|
XM_011517712.2 |
XP_011516014.1
|
7, 8 |
1029 |
|
XM_011517713.2 |
XP_011516015.1
|
6, 7 |
1093 |
|
XM_017014294.1 |
XP_016869783.1
|
5, 6 |
1010 |
|
XM_017014295.1 |
XP_016869784.1
|
6, 7 |
1120 |
|
XM_017014296.1 |
XP_016869785.1
|
5, 6 |
690 |
|
XM_017014297.1 |
XP_016869786.1
|
5, 6 |
686 |
|
GenBank mRNA |
AF394047.1 |
- |
6, 7 |
1211 |
|
AK024996.1 |
- |
4, 5 |
599 |
|
AK093480.1 |
- |
6, 7 |
1155 |
|
AK097653.1 |
- |
6, 7 |
1149 |
|
AL136817.1 |
- |
6, 7 |
1007 |
|
AX748155.1 |
- |
6, 7 |
1155 |
|
BC033786.1 |
- |
5, 6 |
939 |
5166325 |
View Full Product Details