Gene Transcripts

Gene Symbol
FMC1
Entrez Gene ID
154791
Gene Name
formation of mitochondrial complex V assembly factor 1 homolog
Gene Aliases
C7orf55, HSPC268
Chromosome Location
Chr. 7: 139340450 - 139346319 on Build GRCh38
UniGene ID
Hs.718441
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_197964.4 NP_932068.2 1 364
GenBank mRNA AF161386.1 - 1 170
AK093018.1 - 1 133
BC008467.1 - 1 167 4276817
BI117784.1 - 1 364
BM672187.1 - 2 335
CB125801.1 - 1 345

Gene Symbol
C7orf55-LUC7L2
Entrez Gene ID
100996928
Gene Name
C7orf55-LUC7L2 readthrough
Gene Aliases
Chromosome Location
Chr. 7: 139341132 - 139423457 on Build GRCh38
UniGene ID
Hs.731488
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001244584.2 NP_001231513.1 1 364
GenBank mRNA AK297642.1 - 1 136

View Full Product Details