Gene Transcripts

Gene Symbol
PSORS1C1
Entrez Gene ID
170679
Gene Name
psoriasis susceptibility 1 candidate 1
Gene Aliases
C6orf16, SEEK1
Chromosome Location
Chr. 6: 31114831 - 31140092 on Build GRCh38
UniGene ID
Hs.310958
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014068.2 NP_054787.2 6 680
GenBank mRNA AB031479.1 - 6 665
AF484419.1 - 2 286
BC104870.1 - 4 422 8143873
BI825917.1 - 3 342
DB454051.1 - 4 467

View Full Product Details