Gene Transcripts

Gene Symbol
SLC44A5
Entrez Gene ID
204962
Gene Name
solute carrier family 44 member 5
Gene Aliases
CTL5
Chromosome Location
Chr. 1: 75202131 - 75724261 on Build GRCh38
UniGene ID
Hs.654821
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130058.1 NP_001123530.1 14 1149
NM_001320283.1 NP_001307212.1 12 1110
NM_001320285.1 NP_001307214.1 16 1309
NM_001320287.1 NP_001307216.1 15 1242
NM_152697.4 NP_689910.2 14 1149
XM_006710445.3 XP_006710508.1 12 1036
XM_011540986.2 XP_011539288.1 13 1127
XM_011540987.2 XP_011539289.1 15 1204
XM_017000607.1 XP_016856096.1 13 1175
XM_017000608.1 XP_016856097.1 13 1174
XM_017000609.1 XP_016856098.1 16 2493
XM_017000610.1 XP_016856099.1 16 1571
XM_017000611.1 XP_016856100.1 14 1256
GenBank mRNA AK091400.1 - 2 89
AK093170.1 - 14 1151
AK128063.1 - 15 1202
AK296924.1 - 13 1188
AK296941.1 - 12 1009
AK299573.1 - 12 1110
BC028743.2 - 14 1122 4828797
BC034580.1 - 16 1288 4824861
BC051740.1 - 16 1288 4824908

View Full Product Details