Gene Transcripts

Gene Symbol
OR4C11
Entrez Gene ID
219429
Gene Name
olfactory receptor family 4 subfamily C member 11
Gene Aliases
OR11-136, OR4C11P
Chromosome Location
Chr. 11: 55603354 - 55604398 on Build GRCh38
UniGene ID
Hs.553620
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004700.2 NP_001004700.2 1 310
GenBank mRNA BC140727.1 - 1 310

View Full Product Details